Learn More
Abcam IO x 2, HIF-1alpha prolyl hydro x ylase-2 (PHD2) inhibitor, 5MG
Catalog No. 502672824
Encompass_Preferred
MW 352.3 Da, Purity >98%. Potent, cell-permeable, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor (IC₅₀ = 21 nM). Displays over 100-fold selectivity for PHD2 over factor inhibiting HIF-1 (FIH-1) and histone demethylases JMJD2A, JMJD2C, JMJD2E and JMJD3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2824 Supplier Abcam Supplier No. AB2186045MG
MW 352.3 Da, Purity >98%. Potent, cell-permeable, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor (IC₅₀ = 21 nM). Displays over 100-fold selectivity for PHD2 over factor inhibiting HIF-1 (FIH-1) and histone demethylases JMJD2A, JMJD2C, JMJD2E and JMJD3.
The product is subject to the following: Abcam Restricted Use Statement