Learn More
Apexbio Technology LLC IOX2(Glycine) 931398-72-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062368
Encompass_Preferred
IOX2(Glycine) (CAS 931398-72-0) is a selective inhibitor targeting hypoxia-inducible factor-prolyl hydroxylase-2 (PHD2) a key enzyme regulating oxygen sensing by mediating hydroxylation of HIF-1 IOX2 exhibits high target specificity demonstrating an IC50 of approximately 21 nM against PHD2 in vitro and negligible inhibition (over 100-fold selectivity) toward related enzymes such as JMJD2 isoforms and the 2OG-oxygenase FIH In RCC4 cells IOX2 at 50 M effectively suppresses HIF-1 hydroxylation highlighting its utility as a pharmacological probe in studying hypoxia signaling pathways and oxygen-dependent cellular responses
Catalog No. 50-000-62368 Supplier Apexbio Technology LLC Supplier No. A41895.1
IOX2(Glycine) (CAS 931398-72-0) is a selective inhibitor targeting hypoxia-inducible factor-prolyl hydroxylase-2 (PHD2) a key enzyme regulating oxygen sensing by mediating hydroxylation of HIF-1 IOX2 exhibits high target specificity demonstrating an IC50 of approximately 21 nM against PHD2 in vitro and negligible inhibition (over 100-fold selectivity) toward related enzymes such as JMJD2 isoforms and the 2OG-oxygenase FIH In RCC4 cells IOX2 at 50 M effectively suppresses HIF-1 hydroxylation highlighting its utility as a pharmacological probe in studying hypoxia signaling pathways and oxygen-dependent cellular responses