Learn More
TARGETMOL CHEMICALS INC IOX4 25MG
SDP
Catalog No. 502579114
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IOX4 is a potent PHD2 inhibitor (IC50 = 1.6 nM)purity: 98%
Catalog No. 50-257-9114 Supplier TARGETMOL CHEMICALS INC Supplier No. T788025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IOX4 is a potent PHD2 inhibitor (IC50 = 1.6 nM)purity: 98%