Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ioxitalamic acid | 28179-44-4 | MFCD00867942 | 97.0% | 643.94 | C12H11I3N2O5 | 10MG
SDP

Supplier:  Medchemexpress LLC HY12569510MG

Encompass_Preferred

Ioxitalamic acid is an iodinated X-ray contrast medium supplied as a research-grade solid. It is water-soluble, nephrophilic, and highly permeable, suitable for imaging research and analytical applications. The compound has a molecular weight of 643.94 and a reported purity of 97.0%.

  • Water-soluble iodinated contrast agent.
  • Nephrophilic and highly permeable.
  • Solid form; off-white to light yellow appearance.
  • Purity 97.0%.
  • Molecular formula C12H11I3N2O5; molecular weight 643.94.
  • Storage: in solvent -80°C for 6 months and -20°C for 1 month; protect from light and store under inert gas.

Catalog No. 50-002-38927


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.