Learn More
Medchemexpress LLC HY-P7227 50ug Medchemexpress, IP-10/CXCL10, Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP722750UG
Encompass_Preferred
Medchemexpress, HY-P7227 50ug IP-10/CXCL10, Mouse CAS: Recombinant Mouse IP-10/CXCL10 is a member of the CXC chemokine family with pro-inflammatory and anti-angiogenic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4365