Learn More
Cayman Chemical IP ElutIon Buffer 50mL
SDP
Supplier:  Cayman Chemical 60197150 ML
Encompass_Preferred
IP Elution Buffer has been tested and formulated to work exclusively with Cayman’s Protein A/G Coated Plate Immunoprecipitation Kit (Item No. 601970)
Catalog No. 50-289-6839