Learn More
Biocare Medical, LLC intelliPATH Hematoxylin - 20 ml
SDP
Catalog No. 50719801
Encompass_Preferred
IntelliPATH Hematoxylin is water-based and is specially formulated for counterstaining on Biocare intelliPATH Automated Stainer. IntelliPATH Hematoxylin stains nuclei sky-blue and provides high contrast staining for DAB, AEC, Bajoran Purple, Fast Red and Warp Red procedures.
Catalog No. 50-719-801 Supplier Biocare Medical, LLC Supplier No. IPCS5006G20
IntelliPATH Hematoxylin is water-based and is specially formulated for counterstaining on Biocare intelliPATH Automated Stainer. IntelliPATH Hematoxylin stains nuclei sky-blue and provides high contrast staining for DAB, AEC, Bajoran Purple, Fast Red and Warp Red procedures.