Learn More
Leica Biosystems Leica IP C Inkjet Printer for Tissue Cassettes (Configuration 1 - 149IPC00001)
Catalog No. NC2313864
Encompass
The Leica IP C is a modular inkjet cassette printer designed to print durable alphanumeric characters and 2D-Datamatrix barcodes directly onto tissue cassettes, improving traceability and reducing manual labeling in histology workflows.
  • Prints barcode and alphanumeric labels on tissue cassettes for sample tracking and traceability.
  • Ink-jet printing with chemically resistant ink for durability during histology processing.
  • Supports batch printing (15 cassettes/min) and single-cassette printing (~10 seconds per cassette).
  • Integrates with laboratory information systems via a Windows-compliant driver for centralized workflow.
  • Optional accessories include an external cassette magazine holder and automated unload station.
Catalog No. NC2313864 Supplier Leica Biosystems Supplier No. 149IPC00001
Add to Cart
Add to Cart
Request a Quote
The Leica IP C is a modular inkjet cassette printer designed to print durable alphanumeric characters and 2D-Datamatrix barcodes directly onto tissue cassettes, improving traceability and reducing manual labeling in histology workflows.
  • Prints barcode and alphanumeric labels on tissue cassettes for sample tracking and traceability.
  • Ink-jet printing with chemically resistant ink for durability during histology processing.
  • Supports batch printing (15 cassettes/min) and single-cassette printing (~10 seconds per cassette).
  • Integrates with laboratory information systems via a Windows-compliant driver for centralized workflow.
  • Optional accessories include an external cassette magazine holder and automated unload station.