Learn More
Biocare Medical, LLC intelliPATH FLX Peroxidase Blocking Reagent - 100 ml
SDP
Catalog No. NC0988944
Encompass_Preferred
Peroxidazed Blocking Reagent is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations.
Catalog No. NC0988944 Supplier Biocare Medical, LLC Supplier No. IPB5000L
Peroxidazed Blocking Reagent is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations.