Learn More
Eurofins Discoverx ChemiBrite IP1 Prostanoid Receptor Stable Cell Line, 2 vials. (HTS131L)
Catalog No. 502005941
Encompass
The ChemiBrite IP1 Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2) based activation of PTGIR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGIR, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGIR, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5941 Supplier Eurofins Discoverx Supplier No. HTS131L
Add to Cart
Add to Cart
Request a Quote
The ChemiBrite IP1 Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2) based activation of PTGIR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGIR, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGIR, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.