Learn More
Sino Biological IP-10 / CXCL10 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10768MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IP-10 / CXCL10 (rh IP-10 / CXCL10; Catalog#10768-HNAE; NP_001556.2; Val22-Pro98). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7340