Learn More
Enzo Life Sciences IP3 receptor type 1 polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2540100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1829-1848 of human IP3R1. Species reactivity: Human, Rat, Dog. Applications: IHC, WB. Formulation: Liquid. Neat serum containing 0.05% sodium azide.
Catalog No. 50-201-1782