Learn More
MSC Extech PH100 0 to 14 pH, pH Meter Flat Surface pH Meter Probe, Accurate up to 0.01 pH
Catalog No. 501150119
Encompass
Extech PH100 0 to 14 pH, pH Meter Flat Surface pH Meter Probe, Accurate up to 0.01 pH
Catalog No. 50-115-0119 Supplier MSC Supplier No. 79255592
Extech PH100 0 to 14 pH, pH Meter Flat Surface pH Meter Probe, Accurate up to 0.01 pH