Learn More
MSC Mitutoyo Electronic Point Micrometer, Carbide, 0 in. to 1 in., Ratchet Stop, 1 ea
Catalog No. 5000510827
Encompass
This electronic point micrometer is a precision instrument designed for measuring the web thickness of drills, small grooves, keyways, and other hard-to-reach features. It features carbide-tipped spindle and anvil surfaces with a 30° point angle, ensuring durability and precise contact in narrow spaces. The unit offers an accuracy of 0.0001 in. and includes a long-form calibration certificate.
  • Pointed spindle and anvil for precise measurement of hard-to-reach dimensions
  • Carbide-tipped contact surfaces for wear resistance
  • Electronic operation with data output capability
  • Ratchet stop thimble for consistent measuring force
  • IP65 water resistance rating
  • Includes one SR44 silver oxide battery and calibration certificate
Catalog No. 50-005-10827 Supplier MSC Supplier No. 81299489
This electronic point micrometer is a precision instrument designed for measuring the web thickness of drills, small grooves, keyways, and other hard-to-reach features. It features carbide-tipped spindle and anvil surfaces with a 30° point angle, ensuring durability and precise contact in narrow spaces. The unit offers an accuracy of 0.0001 in. and includes a long-form calibration certificate.
  • Pointed spindle and anvil for precise measurement of hard-to-reach dimensions
  • Carbide-tipped contact surfaces for wear resistance
  • Electronic operation with data output capability
  • Ratchet stop thimble for consistent measuring force
  • IP65 water resistance rating
  • Includes one SR44 silver oxide battery and calibration certificate