Learn More
Aviva Systems Biology Corp Ip6k1 Antibody - N-terminal region (ARP62340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ip6k1. It was validated on Western Blot. . Alternate names: 1200016D08Rik, AU022776, Ihpk1, InsP6, Itpk6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-653