Learn More
Medchemexpress LLC IP7e | 500164-74-9 | 99.65% | 390.43 | 1 MG
SDP
Catalog No. 5000343933
Encompass_Preferred
IP7e is a potent, brain-penetrant, and orally active Nurr1 activator with an EC50 value of 3.9 nM. It is intended for research use only and not for patient use.
  • Potent Nurr1 activator.
  • Brain-penetrant.
  • Orally active.
  • Reduces the incidence and severity of experimental autoimmune encephalomyelitis (EAE) in mice with preventive treatment.
  • Attenuates inflammation and neurodegeneration in the spinal cords of EAE mice through an NF-kB pathway-dependent process.
Catalog No. 50-003-43933 Supplier Medchemexpress LLC Supplier No. HY11027450MG
IP7e is a potent, brain-penetrant, and orally active Nurr1 activator with an EC50 value of 3.9 nM. It is intended for research use only and not for patient use.
  • Potent Nurr1 activator.
  • Brain-penetrant.
  • Orally active.
  • Reduces the incidence and severity of experimental autoimmune encephalomyelitis (EAE) in mice with preventive treatment.
  • Attenuates inflammation and neurodegeneration in the spinal cords of EAE mice through an NF-kB pathway-dependent process.