Learn More
Apexbio Technology LLC IPA-3, 5mg. Cas: 42521-82-4 MFCD: MFCD00388917. Non-ATP competitive Pak1 inhibitor
SDP
Supplier:  Apexbio Technology LLC B21695
Encompass_Preferred
IPA-3 is an autoregulatory domain inhibitor of p21-activated kinase (Pak) with IC50 value of 2.5M [1].IPA-3 is a highly selective and non-ATP-competitive inhibitor that targets the autoregulatory mechanism of group I Paks. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5322