Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropyl alcohol, 67-63-0, MFCD00011674, 1 kg
Catalog No. NC1140585
Encompass
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, ≥99.7%, FCC, FG, Synonym: 2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol. Isopropyl alcohol (IPA, 2-propanol, isopropanol) is a water soluble aliphatic alcohol with a sweet odor. It is one of the volatile flavor compound found in Tricholoma matsutake,a fungi used in Japanese dishes for its aromatic flavor. IPA is also a major flavor component in lemon essence.
Catalog No. NC1140585 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2929071KGK
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, ≥99.7%, FCC, FG, Synonym: 2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol. Isopropyl alcohol (IPA, 2-propanol, isopropanol) is a water soluble aliphatic alcohol with a sweet odor. It is one of the volatile flavor compound found in Tricholoma matsutake,a fungi used in Japanese dishes for its aromatic flavor. IPA is also a major flavor component in lemon essence.