Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™

Catalog No. A459200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Poly Bottle
PolyPac™
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A459200 200 L Poly Bottle
A459-500 500 mL Poly Bottle
A459-1 1 L Poly Bottle
A459-4 4 L Poly Bottle
A459-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A459200
  • $7,890.00
    $6,706.65 / Each of 1

    Save $1,183.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88°C
Color Colorless
Density 0.7850g/cm³
Quantity 200 L
Packaging Poly Bottle
Physical Form Liquid
Refractive Index 1.3706 to 1.3722 (at 25°C)
Assay Percent Range 68.0 to 72.0%
Vapor Pressure 33mmHg at 20°C
Synonym isopropanol
Boiling Point 82°C
Molecular Weight (g/mol) 60.10
Formula Weight 60.1
Percent Purity 68.0 to 72.0%
Chemical Name or Material IPA (Isopropyl Alcohol), 70% v/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.