Learn More
Alkali Scientific IPA, Isopropyl Alcohol 99%, 1 Liter
SDP
Catalog No. 5000659658
Encompass_Preferred
Isopropyl Alcohol 99%, also known as IPA, is a high-purity solvent commonly used for laboratory cleaning, surface disinfection, and as a reagent in various chemical applications. This 1-liter bottle is perfect for regular cleaning tasks, ensuring that surfaces, instruments, and glassware are free of contaminants. IPA is a versatile solvent, used in many applications including protein extraction, enzyme inactivation, and DNA/RNA work. Its high evaporation rate and effectiveness in dissolving a wide range of compounds make it an essential tool in labs where maintaining sterile conditions is critical. It is particularly useful in applications requiring fast evaporation with minimal residue.
Catalog No. 50-006-59658 Supplier Alkali Scientific Supplier No. IPA001CP
Isopropyl Alcohol 99%, also known as IPA, is a high-purity solvent commonly used for laboratory cleaning, surface disinfection, and as a reagent in various chemical applications. This 1-liter bottle is perfect for regular cleaning tasks, ensuring that surfaces, instruments, and glassware are free of contaminants. IPA is a versatile solvent, used in many applications including protein extraction, enzyme inactivation, and DNA/RNA work. Its high evaporation rate and effectiveness in dissolving a wide range of compounds make it an essential tool in labs where maintaining sterile conditions is critical. It is particularly useful in applications requiring fast evaporation with minimal residue.