Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ IPA Resistant Cleanroom Marker

Catalog No. 18003117 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-003-117 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-003-117
Add to Cart
Add to Cart

IPA resistant cleanroom marker, black, for use in controlled environments, on metal, glass or poly.

  • Class 10 certified cleanroom Sharpie with IPA-resistant permanent ink.
  • Will mark on wet or abrasive surfaces.
  • A break-resistant clip and durable tip help provide one of the most durable Sharpies for a controlled environment.
  • The anti-roll barrel shape, ribbed grip area, and lanyard loop allow for more confidence in not contaminating the pen from a dropped cap or dropped pen.
  • Manufacture with a chisel tip.
  • Double-bagged in packs of 12.
  • Individual wrap upon request.
  • For use on wet surfaces, glass surfaces, poly bags, rodac plates, and labware.

Specifications

Material Plastic
Color Black
Product Type Cleanroom Sharpie
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.