Learn More
MilliporeSigma IPAK Gard™ 3/5 Pretreatment Cartridge for Hard Water
Catalog No. IPAKGARDH1 Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
IPAKGARDH1 Tap
1 options
Catalog No. IPAKGARDH1 Supplier MilliporeSigma Supplier No. IPAKGARDH1
Get an Extra Lab Water Cartridge with Purchase
View all promotions
Add to Cart
Add to Cart
For Milli-Q™ IQ/IX 7003/05, Milli-Q™ EQ 7008/16 water systems to protect RO membranes. This product replaces IPAKGARH1.
  • Pleated filter removes particulates and colloids, and activated carbon block removes free chlorine from hard feed water to ensure RO membrane performance over years.
  • Polyphosphate beads protect against scaling and precipitates.
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a drain cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 63% of the parts suppliers are ISO 14001 environmental management system certified.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Purification Method Reverse osmosis
Type Pretreatment Cartridge