Learn More
MilliporeSigma IPAK Gard™ 3/5 Pretreatment Cartridge
Catalog No. IPAKGARD1 Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
IPAKGARD1 Tap
1 options
Catalog No. IPAKGARD1 Supplier MilliporeSigma Supplier No. IPAKGARD1
Get an Extra Lab Water Cartridge with Purchase
View all promotions
Add to Cart
Add to Cart
For Milli-Q™ IQ/IX 7003/05 and Milli-Q™ EQ 7008/16 water systems to protect RO membranes. This product replaces IPAKGARA1.
  • Pleated filter removes particulates and colloids, and activated carbon block removes free chlorine from potable water to ensure RO membrane performance over years.
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a drain cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 63% of the parts suppliers are ISO 14001 environmental management system certified.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Purification Method Reverse osmosis
Type Pretreatment Cartridge