Learn More
MilliporeSigma™ IPAK Gard™ 3-5 Pretreatment Pack
Catalog No. IPAKGARA1 Shop All MilliporeSigma Products
Change view
Click to view available options
IPAK Gard™ 3-5 Pretreatment Pack
Catalog No.
IPAKGARA1 IPAK Gard™ 3-5 Pretreatment Pack
1 options
Catalog No. IPAKGARA1 Supplier MilliporeSigma™ Supplier No. IPAKGARA1
For optimal protection of Reverse Osmosis membrane for use with Milli-Q™ IQ 7003 and IQ 7005 systems
High efficiency removal of colloids, particles, free chlorine and minerals for improved system performance.
Benefits:
  • Optimal protection of reverse osmosis (RO) membrane
  • Ensures system lifetime and performance are optimized
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Milli-Q™ IQ 7003 and IQ 7005 Systems