Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IPAK GARD™ Pretreatment Pack for Water Purification Systems

Catalog No. IPAKGARH2 Shop All MilliporeSigma Products
Change view
Click to view available options
Model:
10-15 pack
H 10-15 pack
H 3-5 pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
IPAKGARH2 H 10-15 pack
IPAKGARA2 10-15 pack
IPAKGARH1 H 3-5 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. IPAKGARH2 Supplier MilliporeSigma™ Supplier No. IPAKGARH2

missing translation for 'validMainframeMsg'

Milli-Q™ IQ 7003/05/10/15 Water Purification Systems are integrated ultrapure and pure water systems that are intelligent, easy-to-use and environmentally friendly

TRUSTED_SUSTAINABILITY

Specifications

Capacity Up to 2 L/min.
Flow Rate Up to 2 L/min.
For Use With (Equipment) Milli-Q™ IQ 7003/7005/7010/7015 Systems
Includes Pretreatment pack
Model H 10-15 pack
Electrical Requirements Voltage: 100 to 240 V ±10%; Frequency 50 to 60 Hz ±10%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.