Learn More
MilliporeSigma IPAK Meta™ Polishing Cartridge for Elix™ Purified Water
Catalog No. IPAKMETEX Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Elix Purified Water
Catalog No. Feedwater Source
IPAKMETEX Elix Purified Water
1 options
Catalog No. IPAKMETEX Supplier MilliporeSigma Supplier No. IPAKMETEX
Get an Extra Lab Water Cartridge with Purchase
View all promotions
Add to Cart
Add to Cart
For Milli-Q™ IQ/EQ 7000 ultrapure water systems fed by Elix™ pure water. This product replaces IPAKMETA1.
  • For Milli-Q™ IQ/EQ 7000 ultrapure water purification systems fed by Elix™ water
  • Delivers ultrapure (Type 1) water for sensitive applications
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Elix Purified Water
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Polishing Cartridge