Learn More
MilliporeSigma IPAK Meta™ Polishing Cartridge for Purified Water
Catalog No. IPAKMET00 Shop All MilliporeSigma Products
Change view
Click to view available options
Purification Method:
Ultrapure
Catalog No. Purification Method
IPAKMET00 Ultrapure
1 options
Catalog No. IPAKMET00 Supplier MilliporeSigma Supplier No. IPAKMET00
Add to Cart
Add to Cart
For Milli-Q™ IQ/EQ 7000 and EQ 7008/16 ultrapure water systems fed with purified water. This product replaces IPAKMETA1.
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a drain cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 78% of the parts suppliers are ISO 14001 environmental management system-certified.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Polishing Cartridge