Learn More
Medchemexpress LLC Ipatasertib | 1001264-89-6 | 458.00 | 200 MG
SDP
Catalog No. 5000366052
Encompass_Preferred
Ipatasertib (GDC-0068) is an orally active, highly selective, and ATP-competitive pan-Akt inhibitor. It activates FoxO3a and NF-κB through Akt inhibition, leading to p53-independent activation of PUMA, and induces apoptosis in cancer cells and inhibits tumor growth in xenograft mouse models.
  • Orally active and highly selective pan-Akt inhibitor
  • ATP-competitive inhibition of Akt1, Akt2, and Akt3
  • Synchronously activates FoxO3a and NF-κB
  • Leads to p53-independent activation of PUMA
  • Induces apoptosis in cancer cells
  • Inhibits tumor growth in xenograft mouse models
Catalog No. 50-003-66052 Supplier Medchemexpress LLC Supplier No. 200MGHY15186
Ipatasertib (GDC-0068) is an orally active, highly selective, and ATP-competitive pan-Akt inhibitor. It activates FoxO3a and NF-κB through Akt inhibition, leading to p53-independent activation of PUMA, and induces apoptosis in cancer cells and inhibits tumor growth in xenograft mouse models.
  • Orally active and highly selective pan-Akt inhibitor
  • ATP-competitive inhibition of Akt1, Akt2, and Akt3
  • Synchronously activates FoxO3a and NF-κB
  • Leads to p53-independent activation of PUMA
  • Induces apoptosis in cancer cells
  • Inhibits tumor growth in xenograft mouse models