Learn More
Medchemexpress LLC HY-15186 10mg Medchemexpress, Ipatasertib CAS:1001264-89-6 Purity:>98%
SDP
Catalog No. 502029092
Encompass_Preferred
Medchemexpress, HY-15186 10mg Ipatasertib CAS:1001264-89-6 Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9092 Supplier Medchemexpress LLC Supplier No. HY1518610MG
Medchemexpress, HY-15186 10mg Ipatasertib CAS:1001264-89-6 Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.