Learn More
Selleck Chemical LLC Ipatasertib (GDC-0068) S2808-100mg
SDP
Catalog No. 503111948
Encompass_Preferred
Ipatasertib (GDC-0068 RG7440) is a highly selective pan-Akt inhibitor targeting Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM in cell-free assays 620-fold selectivity over PKA Phase 2
Catalog No. 50-311-1948 Supplier Selleck Chemical LLC Supplier No. S2808100MG
Ipatasertib (GDC-0068 RG7440) is a highly selective pan-Akt inhibitor targeting Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM in cell-free assays 620-fold selectivity over PKA Phase 2