Learn More
TARGETMOL CHEMICALS INC IPATASERTIB 25MG
SDP
Catalog No. 502579472
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ipatasertib (GDC-0068) (GDC-0068) a highly specific pan-Akt inhibitor targets Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM respectively. purity: 99%
Catalog No. 50-257-9472 Supplier TARGETMOL CHEMICALS INC Supplier No. T625225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ipatasertib (GDC-0068) (GDC-0068) a highly specific pan-Akt inhibitor targets Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM respectively. purity: 99%