Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ipatasertib dihydrochloride | 1396257-94-5 | C24H34Cl3N5O2 | 1 ML
SDP

Catalog No. 5000374855
Encompass_Preferred

Ipatasertib dihydrochloride (GDC-0068 dihydrochloride) is a highly selective and ATP-competitive pan-Akt inhibitor. It exhibits IC50s of 5 nM for Akt1, 18 nM for Akt2, and 8 nM for Akt3. This product is intended for research use only.

  • Highly selective and ATP-competitive pan-Akt inhibitor
  • IC50s of 5 nM for Akt1, 18 nM for Akt2, and 8 nM for Akt3
  • Purity 99.19%
  • Molecular weight 530.92
  • Chemical formula C24H34Cl3N5O2
  • Provided as a 10 mM solution in DMSO

Catalog No. 50-003-74855 Supplier Medchemexpress LLC Supplier No. HY15186A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ipatasertib dihydrochloride (GDC-0068 dihydrochloride) is a highly selective and ATP-competitive pan-Akt inhibitor. It exhibits IC50s of 5 nM for Akt1, 18 nM for Akt2, and 8 nM for Akt3. This product is intended for research use only.

  • Highly selective and ATP-competitive pan-Akt inhibitor
  • IC50s of 5 nM for Akt1, 18 nM for Akt2, and 8 nM for Akt3
  • Purity 99.19%
  • Molecular weight 530.92
  • Chemical formula C24H34Cl3N5O2
  • Provided as a 10 mM solution in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.