Learn More
Medchemexpress LLC Ipatasertib dihydrochloride | 1396257-94-5 | C24H34Cl3N5O2 | 100 MG
SDP
Catalog No. 5000372297
Encompass_Preferred
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor. It is provided as a light yellow to yellow solid and is intended for research use only.
  • Highly selective and ATP-competitive pan-Akt inhibitor.
  • Inhibits Akt1, Akt2, and Akt3 with IC50s of 5, 18, and 8 nM, respectively.
  • Demonstrates high selectivity against PKA and p70S6K.
  • Efficacious in cell lines and xenograft models with Akt activation.
  • Can achieve tumor growth delay, stasis, or regression.
  • Enhances tumor growth inhibition in combination therapies.
Catalog No. 50-003-72297 Supplier Medchemexpress LLC Supplier No. HY15186A100MG
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor. It is provided as a light yellow to yellow solid and is intended for research use only.
  • Highly selective and ATP-competitive pan-Akt inhibitor.
  • Inhibits Akt1, Akt2, and Akt3 with IC50s of 5, 18, and 8 nM, respectively.
  • Demonstrates high selectivity against PKA and p70S6K.
  • Efficacious in cell lines and xenograft models with Akt activation.
  • Can achieve tumor growth delay, stasis, or regression.
  • Enhances tumor growth inhibition in combination therapies.