Learn More
Medchemexpress LLC Ipatasertib dihydrochloride | 1396257-94-5 | C24H34Cl3N5O2 | 200 MG
SDP
Catalog No. 5000373900
Encompass_Preferred
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor, with IC50s of 5, 18, and 8 nM for Akt1, Akt2, and Akt3 respectively. It demonstrates potent efficacy in xenograft models where Akt is activated due to genetic alterations, leading to tumor growth delay, stasis, or regression. This compound is intended for research use only.
  • Highly selective and ATP-competitive pan-Akt inhibitor.
  • IC50s of 5, 18, and 8 nM for Akt1, Akt2, and Akt3.
  • Significant selectivity for Akt1 compared to closely related kinases.
  • Effective in xenograft models with Akt activation due to genetic alterations.
  • Induces tumor growth delay, stasis, or regression in vivo.
  • Enhances tumor regression and stasis when used in combination therapies.
  • Increases tumor growth inhibition in ovarian cancer models when combined with other agents.
Catalog No. 50-003-73900 Supplier Medchemexpress LLC Supplier No. HY15186A200MG
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor, with IC50s of 5, 18, and 8 nM for Akt1, Akt2, and Akt3 respectively. It demonstrates potent efficacy in xenograft models where Akt is activated due to genetic alterations, leading to tumor growth delay, stasis, or regression. This compound is intended for research use only.
  • Highly selective and ATP-competitive pan-Akt inhibitor.
  • IC50s of 5, 18, and 8 nM for Akt1, Akt2, and Akt3.
  • Significant selectivity for Akt1 compared to closely related kinases.
  • Effective in xenograft models with Akt activation due to genetic alterations.
  • Induces tumor growth delay, stasis, or regression in vivo.
  • Enhances tumor regression and stasis when used in combination therapies.
  • Increases tumor growth inhibition in ovarian cancer models when combined with other agents.