Learn More
Medchemexpress LLC Ipatasertib dihydrochloride | 1396257-94-5 | C24H34Cl3N5O2 | 500 MG
SDP
Catalog No. 5000374893
Encompass_Preferred
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor. It exhibits significant selectivity for Akt1 over closely related kinases PKA and p70S6K, inhibiting only a few other kinases at higher concentrations.
  • Highly selective and ATP-competitive pan-Akt inhibitor
  • Inhibits Akt1, Akt2, and Akt3
  • Exhibits high selectivity against PKA and p70S6K
  • Inhibits only three other kinases by more than 70% at 1 μM concentration in a panel of 230 protein kinases
Catalog No. 50-003-74893 Supplier Medchemexpress LLC Supplier No. HY15186A500MG
Ipatasertib dihydrochloride is a highly selective and ATP-competitive pan-Akt inhibitor. It exhibits significant selectivity for Akt1 over closely related kinases PKA and p70S6K, inhibiting only a few other kinases at higher concentrations.
  • Highly selective and ATP-competitive pan-Akt inhibitor
  • Inhibits Akt1, Akt2, and Akt3
  • Exhibits high selectivity against PKA and p70S6K
  • Inhibits only three other kinases by more than 70% at 1 μM concentration in a panel of 230 protein kinases