Learn More
Selleck Chemical LLC Ipatasertib (GDC-0068) 10mM/1mL 1001264-89-6 RG7440
SDP
Supplier:  Selleck Chemical LLC S280810MM/1ML
Encompass_Preferred
Ipatasertib (GDC-0068, RG7440) is a highly selective pan-Akt inhibitor targeting Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM in cell-free assays, 620-fold selectivity over PKA. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0742252