Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ipatasertib-NH2 | 1001382-14-4 | 99.6% | 100 MG
SDP

Catalog No. 5000340846
Encompass_Preferred

Ipatasertib-NH2 is a ligand for target protein AKT for PROTAC (INY-03-041). INY-03-041 is composed of Ipatasertib-NH2, a ten-hydrocarbon linker, and a CRBN ligand Lenalidomide for E3 ubiquitin ligase. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.

  • Target: AKT
  • Cellular effect: inhibition of Akt1 in human LNCaP cells with an IC50 of 1454 nM
  • Molecular weight: 415.92
  • Formula: C21H26ClN5O2
  • Appearance: solid
  • Color: white to light yellow

Catalog No. 50-003-40846 Supplier Medchemexpress LLC Supplier No. HY130988100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ipatasertib-NH2 is a ligand for target protein AKT for PROTAC (INY-03-041). INY-03-041 is composed of Ipatasertib-NH2, a ten-hydrocarbon linker, and a CRBN ligand Lenalidomide for E3 ubiquitin ligase. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.

  • Target: AKT
  • Cellular effect: inhibition of Akt1 in human LNCaP cells with an IC50 of 1454 nM
  • Molecular weight: 415.92
  • Formula: C21H26ClN5O2
  • Appearance: solid
  • Color: white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.