Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ipatasertib-NH2 | 1001382-14-4 | 99.6% | 50 MG
SDP

Catalog No. 5000339941
Encompass_Preferred

Ipatasertib-NH2, also known as GDC-0068-NH2 or RG7440-NH2, functions as a ligand for the target protein AKT within PROTAC (INY-03-041). This PROTAC molecule is specifically designed by combining Ipatasertib-NH2 with a ten-hydrocarbon linker and a CRBN ligand, Lenalidomide, to facilitate E3 ubiquitin ligase activity.

  • Purity of 99.6%
  • Molecular weight of 415.92
  • Chemical formula C21H26ClN5O2
  • Appearance as a white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Ligand for target protein AKT in PROTAC research

Catalog No. 50-003-39941 Supplier Medchemexpress LLC Supplier No. HY13098850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ipatasertib-NH2, also known as GDC-0068-NH2 or RG7440-NH2, functions as a ligand for the target protein AKT within PROTAC (INY-03-041). This PROTAC molecule is specifically designed by combining Ipatasertib-NH2 with a ten-hydrocarbon linker and a CRBN ligand, Lenalidomide, to facilitate E3 ubiquitin ligase activity.

  • Purity of 99.6%
  • Molecular weight of 415.92
  • Chemical formula C21H26ClN5O2
  • Appearance as a white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Ligand for target protein AKT in PROTAC research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.