Learn More
Medchemexpress LLC Ipatasertib-NH2 | 1001382-14-4 | 99.6% | C21H26ClN5O2 | 10MG
SDP
Catalog No. 5000261797
Encompass_Preferred
Ipatasertib-NH2 is the amide derivative of the AKT inhibitor ipatasertib, provided as a research-grade ligand for chemical biology and PROTAC development. It is intended for in vitro research use and is supplied with characterization and safety documentation.
  • High purity (≈99.6%) suitable for research applications.
  • Available as a solid and as 10 mM solutions in DMSO for ready use.
  • Molecular formula C21H26ClN5O2; molecular weight 415.92.
  • Used as an AKT-binding ligand in PROTAC constructs and target engagement studies.
  • Supplied with characterization data and an accessible safety data sheet.
Catalog No. 50-002-61797 Supplier Medchemexpress LLC Supplier No. HY13098810MG
Ipatasertib-NH2 is the amide derivative of the AKT inhibitor ipatasertib, provided as a research-grade ligand for chemical biology and PROTAC development. It is intended for in vitro research use and is supplied with characterization and safety documentation.
  • High purity (≈99.6%) suitable for research applications.
  • Available as a solid and as 10 mM solutions in DMSO for ready use.
  • Molecular formula C21H26ClN5O2; molecular weight 415.92.
  • Used as an AKT-binding ligand in PROTAC constructs and target engagement studies.
  • Supplied with characterization data and an accessible safety data sheet.