Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Ipc2B(Allyl)Borane Solution

Catalog No. 67853825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-853-825ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-853-825ML Supplier Sigma Aldrich Supplier No. 67853825ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This allylboration reagent is a convenient, salt-free, stable solution of a chiral allyl borane for asymmetric allylation of aldehydes leading to chiral homoallylic alcohols.Under refrigerator storage, these salt-free, Ipc2B(allyl) reagents do not show any appreciable decrease in selectivity is observed after several months. They exhibit excellent reactivity and reactions can be performed at 5C, even in water.

Specifications

CAS 406680-94-2
Melting Point 109°C to 111°C
Molecular Formula C27H16F5O4P
Linear Formula C27H16F5O4P
MDL Number MFCD09265125
Quantity 100 mg
Synonym 1-Methyl-4-(pentafluorophenyl)-2-(diphenylphosphino)-1,4-benzenedicarboxylate
Molecular Weight (g/mol) 530.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.