Learn More
FUJIFILM Biosciences Ipconazole Standard
Catalog No. 5000450353
Encompass_Preferred
Ipconazole is a fungicide used in agricultural settings This analytical standard provides a reliable reference material for detection and quantification in pesticide residue assays Provided in crystalline solid form and soluble in acetone
Catalog No. 50-004-50353 Supplier FUJIFILM Biosciences Supplier No. 09804221
Ipconazole is a fungicide used in agricultural settings This analytical standard provides a reliable reference material for detection and quantification in pesticide residue assays Provided in crystalline solid form and soluble in acetone