Learn More
Cayman Chemical Iperoxo (iodide), Purity: ≥98%, CAS: 247079-84-1, 10 mg
SDP
Catalog No. 502940294
Encompass_Preferred
An agonist of muscarinic acetylcholine receptors; stimulates [35S]GTPγS binding CHO cell membranes expressing human M2 muscarinic receptors and cell membranes expressing M4 muscarinic receptors (EC50s = 2.12 and 8.47 nM, respectively); induces M1-dependent inhibition of twitch response in electrically-stimulated isolated rabbit vas deferens (pD2 = 9.87) and M3-mediated contraction of isolated guinea pig ileum (pD2 = 9.78); reduces formalin-induced paw licking and acetic acid-induced writhing in mice (ED50s = 0.004 and 0.001 mg/kg, respectively)
Catalog No. 50-294-0294 Supplier Cayman Chemical Supplier No. 2723510 MG
An agonist of muscarinic acetylcholine receptors; stimulates [35S]GTPγS binding CHO cell membranes expressing human M2 muscarinic receptors and cell membranes expressing M4 muscarinic receptors (EC50s = 2.12 and 8.47 nM, respectively); induces M1-dependent inhibition of twitch response in electrically-stimulated isolated rabbit vas deferens (pD2 = 9.87) and M3-mediated contraction of isolated guinea pig ileum (pD2 = 9.78); reduces formalin-induced paw licking and acetic acid-induced writhing in mice (ED50s = 0.004 and 0.001 mg/kg, respectively)