Learn More
FUJIFILM Biosciences Ipfencarbazone Standard
Catalog No. 5000450354
Encompass_Preferred
This standard is used in analytical labs to detect and quantify pesticide residues Provided in crystalline powder form with high purity confirmed by GC and qNMR methods
Catalog No. 50-004-50354 Supplier FUJIFILM Biosciences Supplier No. 09907051
This standard is used in analytical labs to detect and quantify pesticide residues Provided in crystalline powder form with high purity confirmed by GC and qNMR methods