Learn More
Diagnocine LLC iPGell
SDP
Catalog No. NC1701136
Encompass
Making a section of floating cells or 3D cultured cells (such as embryoid body, spheroid, etc) is burdensome work for researchers. Conventional methods are cytospin, smear slide, or agarose gel but these method gives damages to your cell sample as the cells are dried or heated during sample preparaiton. Moreover, for cytospin or smear slide method, you need to start again from sample preparation. iPGell will greatly help you to achieve this just by using two components. Cells can be rapidly jellified with unheated condition. Any toxic components affect to cells are not contained. Jellified sample can be soaked into fixation solution and then you can make a sample block (frozen / paraffin) like the way to make tissue block.
Catalog No. NC1701136 Supplier Diagnocine LLC Supplier No. FNKPG201
Making a section of floating cells or 3D cultured cells (such as embryoid body, spheroid, etc) is burdensome work for researchers. Conventional methods are cytospin, smear slide, or agarose gel but these method gives damages to your cell sample as the cells are dried or heated during sample preparaiton. Moreover, for cytospin or smear slide method, you need to start again from sample preparation. iPGell will greatly help you to achieve this just by using two components. Cells can be rapidly jellified with unheated condition. Any toxic components affect to cells are not contained. Jellified sample can be soaked into fixation solution and then you can make a sample block (frozen / paraffin) like the way to make tissue block.