Learn More
Selleck Chemical LLC Duvelisib (IPI-145) 5mg 1201438-56-3 INK1197
SDP
Catalog No. NC0727626
Encompass_Preferred
Duvelisib (IPI-145, INK1197) is a novel and selective PI3K ?/? inhibitor with Ki and IC50 of 23 pM/243 pM and 1 nM/50 nM in cell-free assays, highly selective for PI3K ?/? than other protein kinases. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0727626 Supplier Selleck Chemical LLC Supplier No. S70285MG
Duvelisib (IPI-145, INK1197) is a novel and selective PI3K ?/? inhibitor with Ki and IC50 of 23 pM/243 pM and 1 nM/50 nM in cell-free assays, highly selective for PI3K ?/? than other protein kinases. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.