Learn More
TARGETMOL CHEMICALS INC IPI-493 1MG
SDP
Catalog No. 502876805
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. IPI-493 (17-amino17demethoxy) is a novel HSP90 inhibitor with antitumor activity. Purity 100%
Catalog No. 50-287-6805 Supplier TARGETMOL CHEMICALS INC Supplier No. T321881MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. IPI-493 (17-amino17demethoxy) is a novel HSP90 inhibitor with antitumor activity. Purity 100%