Learn More
Selleck Chemical LLC IPI-9119-5mg
SDP
Supplier:  Selleck Chemical LLC E26675MG
Encompass_Preferred
IPI-9119 is a potent, selective, and irreversible inhibitor of fatty acid synthase (FASN), which can inhibit the FASN thioesterase domain by promoting acylation of the catalytic serine with an IC50 of 0.3 nM.
Catalog No. 50-238-9809