Learn More
Medchemexpress LLC HY-124628 5mg , IPI-9119 CAS:1346564-56-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246285MG
Encompass_Preferred
HY-124628 5mg IPI-9119 CAS: 1346564-56-4 IPI-9119 is an orally active, selective and irreversible FASN inhibitor with an IC50 of 0.3 nM in vitro biochemical assay. IPI-9119 inhibits tumor growth of castration-resistant prostate cancer (CRPC) xenografts mouse models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1716