Learn More
Medchemexpress LLC HY-100716 100mg Medchemexpress, IPI549 CAS:1693758-51-8 Purity:98%
SDP
Catalog No. 501636808
Encompass_Preferred
Medchemexpress,HY-100716 100mg IPI549 CAS:1693758-51-8 Formula:C30H24N8O2 IPI549 is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6808 Supplier Medchemexpress LLC Supplier No. HY100716100MG
Medchemexpress,HY-100716 100mg IPI549 CAS:1693758-51-8 Formula:C30H24N8O2 IPI549 is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.