Learn More
Medchemexpress LLC HY-100716 50mg Medchemexpress, IPI549 CAS:1693758-51-8 Purity:>98%
SDP
Catalog No. 502028634
Encompass_Preferred
Medchemexpress, HY-100716 50mg IPI549 CAS:1693758-51-8 IPI549 is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. IPI549 shows >100-fold selectivity over other lipid and protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8634 Supplier Medchemexpress LLC Supplier No. HY10071650MG
Medchemexpress, HY-100716 50mg IPI549 CAS:1693758-51-8 IPI549 is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. IPI549 shows >100-fold selectivity over other lipid and protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.